Learn More
CPNE1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of CPNE1.
Supplier: Abnova Corporation PAB19677
Description
Calcium-dependent membrane-binding proteins may regulate molecular events at the interface of the cell membrane and cytoplasm. This gene encodes a calcium-dependent protein that also contains two N-terminal type II C2 domains and an integrin A domain-like sequence in the C-terminus. However, the encoded protein does not contain a predicted signal sequence or transmembrane domains. This protein has a broad tissue distribution and it may function in membrane trafficking. This gene and the gene for RNA binding motif protein 12 overlap at map location 20q11.21. Alternate splicing results in multiple transcript variants encoding different proteins. [provided by RefSeq
Sequence: C-APLKPLPPSAKDPASpecifications
CPNE1 | |
Polyclonal | |
Goat polyclonal antibody raised against synthetic peptide of CPNE1. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_690902.1;NP_003906.2 | |
CPNE1 | |
A synthetic peptide corresponding to C-terminus of human CPNE1. | |
100 μg | |
Primary | |
This antibody is expected to recognize isoform a and b (NP_003906.2; NP_690902.1). Reported variants represent identical protein: NP_690905.1, NP_690902.1, NP_690903.1, NP_690906.1, NP_690904.1, NP_690907.1, NP_690908.1. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:16000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user. | |
CPNE1 | |
COPN1/CPN1/MGC1142 | |
Goat | |
Antigen affinity purification | |
RUO | |
8904 | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only