Learn More
CPT2 Goat anti-Human, Mouse, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of CPT2.
Supplier: Abnova Corporation PAB18741
Description
The protein encoded by this gene is a nuclear protein which is transported to the mitochondrial inner membrane. Together with carnitine palmitoyltransferase I, the encoded protein oxidizes long-chain fatty acids in the mitochondria. Defects in this gene are associated with mitochondrial long-chain fatty-acid (LCFA) oxidation disorders. [provided by RefSeq
Sequence: C-PKSEYNDQLTRATNSpecifications
CPT2 | |
Polyclonal | |
Unconjugated | |
ELISA (1:8000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
CPT2 | |
CPT1/CPTASE | |
Goat | |
Antigen affinity purification | |
RUO | |
1376 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of CPT2. | |
In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
NP_000089.1 | |
CPT2 | |
A synthetic peptide corresponding to amino acids 141-154 at internal region of human CPT2. | |
100 μg | |
Primary | |
Human, Mouse | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only