Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CREM, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89132609 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-609 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-609 Supplier Abnova Corporation Supplier No. PAB6696
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of CREM.

This gene encodes a bZIP transcription factor that binds to the cAMP responsive element found in many viral and cellular promoters. It is an important component of cAMP-mediated signal transduction during the spermatogenetic cycle, as well as other complex processes. Alternative promoter and translation initiation site usage allows this gene to exert spatial and temporal specificity to cAMP responsiveness. Multiple alternatively spliced transcript variants encoding several different isoforms have been found for this gene, with some of them functioning as activators and some as repressors of transcription. [provided by RefSeq

Sequence: LETLKDICSPKTDY

Specifications

Antigen CREM
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of CREM.
Dilution ELISA (1:16000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene CREM
Gene Accession No. NP_853549.1;NP_874387.1;NP_874388.1;NP_874393.1;NP_874394.1;NP_877570.1;NP_877571.1;NP_877572.1;NP_877573.1;NP_878270.1;NP_898883.1
Gene Alias ICER/MGC111110/MGC17881/MGC41893/hCREM-2
Gene Symbols CREM
Host Species Goat
Immunogen A synthetic peptide corresponding to human CREM.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1390
Test Specificity This antibody is expected to recognize the following protein isoforms of human CREM protein: a (NP_853549.1), e (NP_874387.1), f (NP_874388.1), k (NP_874393.1), l (NP_874394.1), m (NP_877570.1), n (NP_877571.1), o (NP_877572.1), p (NP_877573.1), q (NP_878270.1), v (NP_898883.1). This antibody is not expected to cross-react with CREB protein.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.