Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRP Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against native CRP.
Supplier: Abnova Corporation PAB7943
Specifications
CRP | |
Polyclonal | |
Goat polyclonal antibody raised against native CRP. | |
In 15mM KPO4 buffer, 0.85% NaCl, pH 7.4 (0.09% sodium azide) | |
MGC149895/MGC88244/PTX1 | |
Goat | |
1 mg | |
Primary | |
This antibody is specific to human CRP. | |
Store at 4°C. |
ELISA | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
CRP | |
CRP | |
Native purified human CRP. | |
RUO | |
1401 | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction