Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Crtc2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of Crtc2.
Supplier: Abnova Corporation PAB27597
Description
Sequence: HGQQPYHRPINDSpecifications
Crtc2 | |
Polyclonal | |
Unconjugated | |
ELISA (1:32000) Western Blot (0.03-0.1ug/ml) The optimal working dilution should be determined by the end user. | |
Crtc2 | |
4632407F12Rik/Torc2/mTORC2 | |
Goat | |
Affinity Purified | |
RUO | |
74343 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of Crtc2. | |
In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
NP_859066.1 | |
Crtc2 | |
A synthetic peptide corresponding to internal region of mouse Crtc2. | |
100 μg | |
Primary | |
Mouse, Rat |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction