Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CSMD3, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of CSMD3.
Supplier: Abnova Corporation PAB7540
Description
Sequence: KQRTAPKTQYTGCSpecifications
CSMD3 | |
Polyclonal | |
Unconjugated | |
ELISA (1:16000) The optimal working dilution should be determined by the end user. | |
CSMD3 | |
KIAA1894 | |
Goat | |
Antigen affinity purification | |
RUO | |
114788 | |
Human | |
Liquid |
ELISA | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of CSMD3. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_937756.1;NP_937757.1;NP_443132.2 | |
CSMD3 | |
A synthetic peptide corresponding to human CSMD3. | |
100 μg | |
Primary | |
This antibody is expected to recognize all reported isoforms according to NP_937756.1; NP_937757.1 and NP_443132.2. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction