Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTDSP1 Goat, Polyclonal Antibody, Abnova™

Catalog No. 89133086 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-133-086 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-133-086 Supplier Abnova Corporation Supplier No. PAB7256
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of CTDSP1.

Sequence: C-RGKGDQKSAASQK

Specifications

Antigen CTDSP1
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of CTDSP1.
Dilution ELISA (1:32000) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene CTDSP1
Gene Accession No. NP_067021.1;NP_872580.1
Gene Alias NLIIF/SCP1
Gene Symbols CTDSP1
Host Species Goat
Immunogen A synthetic peptide corresponding to human CTDSP1.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58190
Test Specificity This antibody is expected to recognize both reported isoforms (NP_067021.1and NP_872580.1).
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.