Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYB5, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89137651 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-651 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-651 Supplier Abnova Corporation Supplier No. PAB13663
Add to Cart
Add to Cart

Goat polyclonal antibody raised against CYB5.

Specifications

Antigen CYB5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against CYB5.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.05% sodium azide
Host Species Goat
Immunogen Rabbit cytochrome b5 (CYB5).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This is a goat polyclonal antibody and is specific to cytochrome b5 in rabbit but cross reacts with human and rat. This antibody is non-cross reactive with the 50 and 60 KDa species of cytochrome b5.
Target Species Human, Rabbit, Rat
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.