Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyb5r3 Goat anti-Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89121255 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-255 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-255 Supplier Abnova Corporation Supplier No. PAB19727
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of Cyb5r3.

Sequence: CLPNLERVGHPKER

Specifications

Antigen Cyb5r3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of Cyb5r3.
Dilution ELISA (1:16000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene Cyb5r3
Gene Accession No. NP_084063.1
Gene Alias 0610016L08Rik/2500002N19Rik/C85115/Dia-1/Dia1/WU:AL591952.1-001/WU:AL591952.1-002/WU:AL591952.1-003/WU:Cyb5r3
Gene Symbols Cyb5r3
Host Species Goat
Immunogen A synthetic peptide corresponding to C-terminus of mouse Cyb5r3.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109754
Target Species Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.