Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP24A1, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89120527 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-527 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-527 Supplier Abnova Corporation Supplier No. PAB18975
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of CYP24A1.

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This mitochondrial protein initiates the degradation of 1,25-dihydroxyvitamin D3, the physiologically active form of vitamin D3, by hydroxylation of the side chain. In regulating the level of vitamin D3, this enzyme plays a role in calcium homeostasis and the vitamin D endocrine system. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: C-RKYDIQATDN

Specifications

Antigen CYP24A1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of CYP24A1.
Dilution ELISA (1:32000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 0.5mg/mL in Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene CYP24A1
Gene Accession No. NP_000773.2;NP_001122387.1
Gene Alias CP24/CYP24/MGC126273/MGC126274/P450-CC24
Gene Symbols CYP24A1
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids at internal region of human CYP24A1.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1591
Test Specificity This antibody is expected to recognize both reported isoforms (NP_000773.2; NP_001122387.1).
Target Species Human, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.