Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP2C8, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89126305 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-126-305 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-126-305 Supplier Abnova Corporation Supplier No. PAB24588
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of CYP2C8.

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and its expression is induced by phenobarbital. The enzyme is known to metabolize many xenobiotics, including the anticonvulsive drug mephenytoin, benzo(a)pyrene, 7-ethyoxycoumarin, and the anti-cancer drug taxol. This gene is located within a cluster of cytochrome P450 genes on chromosome 10q24. [provided by RefSeq

Sequence: C-KNVALTRSYIREK

Specifications

Antigen CYP2C8
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of CYP2C8.
Dilution ELISA (1:32000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
Gene CYP2C8
Gene Accession No. NP_000761.3;NP_001185782.1;NP_001185783.1
Gene Alias CPC8/MP-12/MP-20
Gene Symbols CYP2C8
Host Species Goat
Immunogen A synthetic peptide corresponding to internal region of human CYP2C8.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1558
Test Specificity This antibody is expected to recognize all reported isoforms (NP_000761.3; NP_001185782.1; NP_001185783.1). No cross-reactivity with other 2C proteins expected. Reported variants represent identical protein: NP_001185782.1, NP_001185784.1
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.