Learn More
CYTH2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of CYTH2.
Supplier: Abnova Corporation PAB7405
Description
The protein encoded by this gene is a member of the PSCD family. Members of this family have identical structural organization that consists of an N-terminal coiled-coil motif, a central Sec7 domain, and a C-terminal pleckstrin homology (PH) domain. The coiled-coil motif is involved in homodimerization, the Sec7 domain contains guanine-nucleotide exchange protein (GEP) activity, and the PH domain interacts with phospholipids and is responsible for association of PSCDs with membranes. Members of this family appear to mediate the regulation of protein sorting and membrane trafficking. The encoded protein exhibits GEP activity in vitro with ARF1, ARF3, and ARF6 and is 83% homologous to CYTH1. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: EDGVYEPPDLTP-CSpecifications
CYTH2 | |
Polyclonal | |
Unconjugated | |
ELISA (1:32000) Western Blot (0.5-1.5 ug/mL) The optimal working dilution should be determined by the end user. | |
CYTH2 | |
ARNO/CTS18/CTS18.1/PSCD2/PSCD2L/SEC7L/Sec7p-L/Sec7p-like | |
Goat | |
Antigen affinity purification | |
RUO | |
9266 | |
Human, Rat | |
Liquid |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of CYTH2. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_004219.2;NP_059431.1 | |
CYTH2 | |
A synthetic peptide corresponding to human CYTH2. | |
100 μg | |
Primary | |
This antibody is expected to recognize both reported isoforms (NP_004219.2, NP_059431.1). | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only