Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D130043K22Rik, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89115174 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-174 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-174 Supplier Abnova Corporation Supplier No. PAB11551
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of D130043K22Rik.

Sequence: C-QGKIKQENKPTLH

Specifications

Antigen D130043K22Rik
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of D130043K22Rik.
Dilution ELISA (1:128000) Western Blot (0.5-1.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene D130043K22Rik
Gene Accession No. NP_001074520.1
Gene Alias 4930451E12Rik
Gene Symbols D130043K22Rik
Host Species Goat
Immunogen A synthetic peptide corresponding to mouse D130043K22Rik.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 210108
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.