Learn More
DACH1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of DACH1.
Supplier: Abnova Corporation PAB18681
Description
This gene encodes a chromatin-associated protein that associates with other DNA-binding transcription factors to regulate gene expression and cell fate determination during development. The protein contains a Ski domain that is highly conserved from Drosophila to human. Expression of this gene is lost in some forms of metastatic cancer, and is correlated with poor prognosis. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: SPVENTPQNNECKSpecifications
DACH1 | |
Polyclonal | |
Unconjugated | |
ELISA (1:64000) Western Blot (0.2-0.6 ug/mL) The optimal working dilution should be determined by the end user. | |
DACH1 | |
DACH/FLJ10138 | |
Goat | |
Antigen affinity purification | |
RUO | |
1602 | |
Human | |
Liquid |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of DACH1. | |
In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
NP_542937.1;NP_542938.1;NP_004383.2 | |
DACH1 | |
A synthetic peptide corresponding to amino acids at internal region of human DACH1. | |
100 μg | |
Primary | |
This antibody is expected to recognize all three reported isoforms(NP_542937.1; NP_542938.1; NP_004383.2). | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only