Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAPP1 Goat anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89120211 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-211 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-211 Supplier Abnova Corporation Supplier No. PAB18652
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of DAPP1.

Sequence: SALCISPEEKTDHK

Specifications

Antigen DAPP1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of DAPP1.
Dilution ELISA (1:4000) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
Gene DAPP1
Gene Accession No. NP_055210.1.
Gene Alias BAM32/DKFZp667E0716
Gene Symbols DAPP1
Host Species Goat
Immunogen A synthetic peptide corresponding to C-terminus of human DAPP1.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27071
Test Specificity This antibody was raised against the C-terminus of NP_055210.1. However, NCBI has changed the version to NP_055210.2 thus losing the C-terminal epitope.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.