Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Goat anti-Digoxin, Polyclonal, Abnova™
Supplier: Abnova Corporation PAB13936
Specifications
| Digoxin | |
| Polyclonal | |
| In antiserum | |
| Digoxin conjugated with BSA. | |
| Primary | |
| Antisera |
| Radioimmune Assays (RIA) | |
| Unconjugated | |
| Goat | |
| 1 mL | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction