Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIXDC1, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89119520 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-119-520 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-119-520 Supplier Abnova Corporation Supplier No. PAB17857
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of DIXDC1.

Sequence: C-DVSYHQVDLERE

Specifications

Antigen DIXDC1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of DIXDC1.
Dilution ELISA (1:64000) Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
Gene DIXDC1
Gene Accession No. NP_001033043.1;NP_219493.1
Gene Alias CCD1/KIAA1735
Gene Symbols DIXDC1
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids at internal region of human DIXDC1.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85458
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.