Learn More
DLC1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of DLC1.
Supplier: Abnova Corporation PAB6661
Description
This gene is deleted in the primary tumor of hepatocellular carcinoma. It maps to 8p22-p21.3, a region frequently deleted in solid tumors. It is suggested that this gene is a candidate tumor suppressor gene for human liver cancer, as well as for prostate, lung, colorectal, and breast cancers. Alternative splicing at this locus results in several transcript variants encoding different isoforms. [provided by RefSeq
Sequence: SVAIRKRSWEEHCSpecifications
DLC1 | |
Polyclonal | |
Unconjugated | |
ELISA (1:32000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) The optimal working dilution should be determined by the end user. | |
DLC1 | |
ARHGAP7/FLJ21120/HP/STARD12/p122-RhoGAP | |
Goat | |
Antigen affinity purification | |
RUO | |
10395 | |
Human | |
Liquid |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of DLC1. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_872584.2;NP_079043.3 | |
DLC1 | |
A synthetic peptide corresponding to N-terminus of human DLC1. | |
100 μg | |
Primary | |
This antibody is expected to recognize isoforms 1 and 3 ( NP_872584.1 and NP_079043.2 resp) but NOT isoform 2 (NP_006085.2). | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only