Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DLG4, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89121184 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-184 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-184 Supplier Abnova Corporation Supplier No. PAB19658
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of DLG4.

This gene encodes a member of the membrane-associated guanylate kinase (MAGUK) family. It heteromultimerizes with another MAGUK protein, DLG2, and is recruited into NMDA receptor and potassium channel clusters. These two MAGUK proteins may interact at postsynaptic sites to form a multimeric scaffold for the clustering of receptors, ion channels, and associated signaling proteins. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: C-EEQARKAFDRATK

Specifications

Antigen DLG4
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of DLG4.
Dilution ELISA (1:8000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene DLG4
Gene Accession No. NP_001356.1;NP_001122299.1
Gene Alias FLJ97752/FLJ98574/PSD95/SAP-90/SAP90
Gene Symbols DLG4
Host Species Goat
Immunogen A synthetic peptide corresponding to C-terminus of human DLG4.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1742
Test Specificity This antibody is expected to recognize both reported isoforms (NP_001356.1; NP_001122299.1)
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.