Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DLL1, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89116109 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-109 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-109 Supplier Abnova Corporation Supplier No. PAB12816
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of DLL1.

DLL1 is a human homolog of the Notch Delta ligand and is a member of the delta/serrate/jagged family. It plays a role in mediating cell fate decisions during hematopoiesis. It may play a role in cell-to-cell communication. [provided by RefSeq

Sequence: C-ATQRHLTVGEEWSQD

Specifications

Antigen DLL1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of DLL1.
Dilution ELISA (1:16000) Western blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene DLL1
Gene Accession No. NP_005609.3
Gene Alias DELTA1/DL1/Delta
Gene Symbols DLL1
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids at internal region of human DLL1.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28514
Test Specificity 75 KDa band observed in Mouse Heart and Rat Heart lysates (calculated MW of 78.1 KDa according to human NP_005609.3, 78.5K Da according to Mouse NP_031891.2 and 77.4 KDa according to Rat NP_114452.1). Recommended concentration: 0.3-1 ug/ml.
Target Species Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.