Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNP, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89137633 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-633 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-633 Supplier Abnova Corporation Supplier No. PAB13594
Add to Cart
Add to Cart

Goat polyclonal antibody raised against DNP.

Specifications

Antigen DNP
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against DNP.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.01% sodium azide)
Host Species Goat
Immunogen DNP conjugated with KLH.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This goat polyclonal antibody is specific to DNP and is intended for use in experiments related to receptor-mediated endocytosis and oxidative stress.
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.