Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EIF3E, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of EIF3E.
Supplier: Abnova Corporation PAB18954
Description
Sequence: C-KLNQNSRSEAPNSpecifications
EIF3E | |
Polyclonal | |
Unconjugated | |
ELISA (1:2000) Western Blot (0.03-0.1 ug/mL) The optimal working dilution should be determined by the end user. | |
EIF3E | |
EIF3-P48/EIF3S6/INT6/eIF3-p46 | |
Goat | |
Antigen affinity purification | |
RUO | |
3646 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of EIF3E. | |
In 0.5mg/mL in Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_001559.1 | |
EIF3E | |
A synthetic peptide corresponding to amino acids at C-terminus of humanEIF3E. | |
100 μg | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction