Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ELF5, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89132230 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-230 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-230 Supplier Abnova Corporation Supplier No. PAB6200
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of ELF5.

The protein encoded by this gene is a member of an epithelium-specific subclass of the Ets transcritpion factor family. In addition to its role in regulating the later stages of terminal differentiation of keratinocytes, it appears to regulate a number of epithelium-specific genes found in tissues containing glandular epithelium such as salivary gland and prostate. It has very low affinity to DNA due to its negative regulatory domain at the amino terminus. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq

Sequence: C-GKNAHGWQEDKL

Specifications

Antigen ELF5
Applications ELISA, Immunohistochemistry (PFA fixed)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of ELF5.
Dilution ELISA (1:128000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-8 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene ELF5
Gene Accession No. NP_938195.1;NP_001413.1
Gene Alias ESE2
Gene Symbols ELF5
Host Species Goat
Immunogen A synthetic peptide corresponding to C-terminus of human ELF5.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2001
Test Specificity This antibody is expected to recognize both reported human isoforms represented by NP_938195.1 and NP_001413.1.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.