Learn More
ERP29, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of ERP29.
Supplier: Abnova Corporation PAB11430
Description
This gene encodes a reticuloplasmin, a protein which resides in the lumen of the endoplasmic reticulum (ER). The protein shows sequence similarity to the protein disulfide isomerase family. However, it lacks the thioredoxin motif characteristic of this family, suggesting that this protein does not function as a disulfide isomerase. The protein dimerizes and is thought to play a role in the processing of secretory proteins within the ER. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq
Sequence: C-EKNKMSDGKKEELQSpecifications
| ERP29 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:2000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user. | |
| ERP29 | |
| C12orf8/ERp28/ERp31/PDI-DB | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 10961 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of ERP29. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_006808.1 | |
| ERP29 | |
| A synthetic peptide corresponding to human ERP29. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only