Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EXOC7, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89132881 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-881 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-881 Supplier Abnova Corporation Supplier No. PAB7019
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of EXOC7.

EXOC7 is a component of the exocyst, which is an evolutionarily conserved octameric protein complex essential for exocytosis. The exocyst targets secretory vesicles at specific domains of the plasma membrane for cell surface expansion and protein secretion (Zuo et al., 2006 [PubMed 17086175]).[supplied by OMIM

Sequence: C-DNIKNDPDKEYN

Specifications

Antigen EXOC7
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of EXOC7.
Dilution ELISA (1:16000) Western blot (0.5-1.5 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene EXOC7
Gene Accession No. NP_001013861.1;NP_0560.34.2
Gene Alias 2-5-3p/DKFZp686J04253/EX070/EXO70/EXOC1/Exo70p/FLJ40965/FLJ46415/YJL085W
Gene Symbols EXOC7
Host Species Goat
Immunogen A synthetic peptide corresponding to human EXOC7.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23265
Test Specificity This antibody is expected to recognize both reported isoforms (NP_001013861.1 and NP_0560.34.2).
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.