Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAM62A, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of FAM62A.
Supplier: Abnova Corporation PAB7548
Description
Sequence: C-QKKRTLSPEFNERSpecifications
FAM62A | |
Polyclonal | |
Unconjugated | |
ELISA (1:8000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
FAM62A | |
KIAA0747/MBC2 | |
Goat | |
Antigen affinity purification | |
RUO | |
23344 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of FAM62A. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_056107.1 | |
FAM62A | |
A synthetic peptide corresponding to human FAM62A. | |
100 μg | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction