Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FBXL11, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89129100 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-100 Supplier Abnova Corporation Supplier No. PAB27589
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of FBXL11.

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbls class and, in addition to an F-box, contains at least 6 highly degenerated leucine-rich repeats. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: PRKDRQVHLTHFELE

Specifications

Antigen FBXL11
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of FBXL11.
Dilution ELISA (1:32000) Western Blot (0.5-2ug/ml) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
Gene FBXL11
Gene Accession No. NP_036440.1
Gene Alias CXXC8/DKFZp434M1735/FBL11/FBL7/FLJ00115/FLJ46431/JHDM1A/KDM2A/KIAA1004/LILINA
Gene Symbols FBXL11
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids 445-459 at internal region of human FBXL11.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22992
Target Species Canine, Chicken, Human, Mouse, Pig, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.