Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FEM1A, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised aganist synthetic peptide of FEM1A.
Supplier: Abnova Corporation PAB15628
Description
Sequence: C-GFIPEDLEAFIELHSpecifications
FEM1A | |
Polyclonal | |
Unconjugated | |
ELISA (1:4000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
FEM1A | |
DKFZp762M136/EPRAP | |
Goat | |
Antigen affinity purification | |
RUO | |
55527 | |
Human | |
Liquid |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised aganist synthetic peptide of FEM1A. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_061178.1 | |
FEM1A | |
A synthetic peptide corresponding to human FEM1A. | |
100 μg | |
Primary | |
Approximately 73.6 KDa band observed in human heart lysates (calculated MW of 73.6 KDa according to NP_061178.1). | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction