Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FH, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89133134 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-133-134 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-133-134 Supplier Abnova Corporation Supplier No. PAB7310
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of FH.

The protein encoded by this gene is an enzymatic component of the tricarboxylic acid (TCA) cycle, or Krebs cycle, and catalyzes the formation of L-malate from fumarate. It exists in both a cytosolic form and an N-terminal extended form, differing only in the translation start site used. The N-terminal extended form is targeted to the mitochondrion, where the removal of the extension generates the same form as in the cytoplasm. It is similar to some thermostable class II fumarases and functions as a homotetramer. Mutations in this gene can cause fumarase deficiency and lead to progressive encephalopathy. [provided by RefSeq

Sequence: C-HPNDHVNKSQSSND

Specifications

Antigen FH
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of FH.
Dilution ELISA (1:64000) Western Blot (0.01-0.03 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene FH
Gene Accession No. NP_000134.2
Gene Alias HLRCC/LRCC/MCL/MCUL1
Gene Symbols FH
Host Species Goat
Immunogen A synthetic peptide corresponding to human FH.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2271
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.