Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FHL1 Goat anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89132599 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-599 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-599 Supplier Abnova Corporation Supplier No. PAB6686
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of FHL1.

LIM proteins, named for 'LIN11, ISL1, and MEC3,' are defined by the possession of a highly conserved double zinc finger motif called the LIM domain.[supplied by OMIM

Sequence: CNKRFVFHNEQVY

Specifications

Antigen FHL1
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of FHL1.
Dilution ELISA (1:32000) Western blot (0.05-0.3 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.2-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene FHL1
Gene Accession No. NP_001440.1
Gene Alias FHL1B/FLH1A/KYO-T/MGC111107/SLIM1/XMPMA/bA535K18.1
Gene Symbols FHL1
Host Species Goat
Immunogen A synthetic peptide corresponding to internal region of human FHL1.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2273
Test Specificity This antibody is expected to recognize human FHL1 protein.
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.