Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FHL2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of FHL2.
Supplier: Abnova Corporation PAB6596
Description
LIM proteins contain a highly conserved double zinc finger motif called the LIM domain.[supplied by OMIM
Sequence: RDDILCPDCGKDISpecifications
FHL2 | |
Polyclonal | |
Unconjugated | |
ELISA (1:16000) Western blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
FHL2 | |
AAG11/DRAL/SLIM3 | |
Goat | |
Antigen affinity purification | |
RUO | |
2274 | |
Human, Mouse | |
Liquid |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of FHL2. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_001441.4;NP_963849.1;NP_963851.2;NP_0010.34581.1 | |
FHL2 | |
A synthetic peptide corresponding to human FHL2. | |
100 μg | |
Primary | |
NP_001441.4, NP_963849.1, NP_963851.2 and NP_0010.34581.1 represent varients of the same protein. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction