Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXG1, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89132262 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-262 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-262 Supplier Abnova Corporation Supplier No. PAB6235
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of FOXG1.

This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of the brain and telencephalon. [provided by RefSeq

Sequence: C-HQNQGSSSNPLIH

Specifications

Antigen FOXG1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of FOXG1.
Dilution ELISA (1:64000) Western blot (0.3-1.0 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene FOXG1
Gene Accession No. CAA550.38.1(FOXG1A);NP_005240.3(FOXG1B)
Gene Alias BF1/BF2/FHKL3/FKH2/FKHL1/FKHL2/FKHL3/FKHL4/FOXG1A/FOXG1B/FOXG1C/HBF-1/HBF-2/HBF-3/HBF-G2/HBF2/HFK1/HFK2/HFK3/KHL2/QIN
Gene Symbols FOXG1
Host Species Goat
Immunogen A synthetic peptide corresponding to human FOXG1.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2290
Test Specificity This antibody recognizes both human FOXG1A and FOXG1B, which are almost completely identical.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.