Learn More
FOXK2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of FOXK2.
Supplier: Abnova Corporation PAB6446
Description
The protein encoded by this gene contains a fork head DNA binding domain. This protein can bind to the purine-rich motifs of the HIV long terminal repeat (LTR), and to the similar purine-rich motif in the interleukin 2 (IL2) promoter. It may be involved in the regulation of viral and cellular promoter elements. [provided by RefSeq
Sequence: C-TPPAAVREKGVQNSpecifications
FOXK2 | |
Polyclonal | |
Unconjugated | |
ELISA (1:32000) Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
FOXK2 | |
ILF/ILF-1/ILF1 | |
Goat | |
Antigen affinity purification | |
RUO | |
3607 | |
Human | |
Liquid |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of FOXK2. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_004505 | |
FOXK2 | |
A synthetic peptide corresponding to human FOXK2. | |
100 μg | |
Primary | |
This antibody is expected to recognize isoform 1 (represented by NP_004505) but not isoforms 2 and 3 (represented by NP_852095 and NP_852096) | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only