Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FTCD Goat anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89132578 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-578 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-578 Supplier Abnova Corporation Supplier No. PAB6664
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of FTCD.

FTCD is a bifunctional enzyme that channels 1-carbon units from formiminoglutamate, a metabolite of the histidine degradation pathway, to the folate pool.[supplied by OMIM

Sequence: SQLVECVPNFSEGKNQ

Specifications

Antigen FTCD
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of FTCD.
Dilution ELISA (1:64000) Western blot (0.03-0.5 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (3-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene FTCD
Gene Accession No. NP_006648;NP_996848
Gene Alias LCHC1
Gene Symbols FTCD
Host Species Goat
Immunogen A synthetic peptide corresponding to human FTCD.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10841
Test Specificity This antibody is expected to recognize both reported isoforms (as represented by NP_006648 and NP_996848).
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.