Learn More
FZD9, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of FZD9.
Supplier: Abnova Corporation PAB7463
Description
Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD9 gene is located within the Williams syndrome common deletion region of chromosome 7, and heterozygous deletion of the FZD9 gene may contribute to the Williams syndrome phenotype. FZD9 is expressed predominantly in brain, testis, eye, skeletal muscle, and kidney. [provided by RefSeq]
Sequence: C-TKTDPSLENPTHSpecifications
FZD9 | |
Polyclonal | |
Unconjugated | |
ELISA (1:32000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5-10 ug/mL) The optimal working dilution should be determined by the end user. | |
FZD9 | |
CD349/FZD3 | |
Goat | |
Antigen affinity purification | |
RUO | |
8326 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry (PFA fixed) | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of FZD9. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_0.03499.1 | |
FZD9 | |
A synthetic peptide corresponding to C-terminus of human FZD9. | |
100 μg | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only