Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Goat anti-Glucose-6-Phosphate Dehydrogenase, HRP, Polyclonal, Abnova™

Catalog No. 89137877 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-877 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-877 Supplier Abnova Corporation Supplier No. PAB14244

Specifications

Antigen Glucose-6-Phosphate Dehydrogenase
Applications ELISA
Classification Polyclonal
Conjugate HRP
Formulation Lyophilized from 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (10mg/mL BSA, 0.01% gentamicin sulfate)
Host Species Goat
Immunogen Native Glucose-6-Phosphate Dehydrogenase from yeast.
Quantity 20 mg
Primary or Secondary Primary
Target Species Yeast
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.