Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNB2L1, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89121220 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-220 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-220 Supplier Abnova Corporation Supplier No. PAB19693
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of GNB2L1.

Sequence: NTLGVCKYTVQDESH

Specifications

Antigen GNB2L1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of GNB2L1.
Dilution Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene GNB2L1
Gene Accession No. NP_006089.1
Gene Alias Gnb2-rs1/H12.3/HLC-7/PIG21/RACK1
Gene Symbols GNB2L1
Host Species Goat
Immunogen A synthetic peptide corresponding to internal region of human GNB2L1.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10399
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.