Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR83 Goat anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89126296 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-126-296 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-126-296 Supplier Abnova Corporation Supplier No. PAB24579
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of GPR83.

Sequence: C-RRYGAESQNPTVK

Specifications

Antigen GPR83
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of GPR83.
Dilution ELISA (1:8000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
Gene GPR83
Gene Accession No. NP_057624.3
Gene Alias GIR/GPR72
Gene Symbols GPR83
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids 59-71 at internal region of human GPR83.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10888
Test Specificity The immunizing peptide represents part of an extracellular domain.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.