Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gpx1 Goat anti-Chicken, Mouse, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89129072 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-129-072 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-129-072 Supplier Abnova Corporation Supplier No. PAB27561
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of Gpx1.

Sequence: DYTEMNDLQKRLGPR

Specifications

Antigen Gpx1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of Gpx1.
Dilution ELISA (1:128000) Western Blot (0.1-0.3ug/ml) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
Gene Gpx1
Gene Accession No. NP_032186.2
Gene Alias AI195024/AL033363/CGPx/GPx-1/GSHPx-1/Gpx
Gene Symbols Gpx1
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids 53-67 at internal region of mouse Gpx1.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14775
Target Species Chicken, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.