Learn More
GRIN2B, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of GRIN2B.
Supplier: Abnova Corporation PAB7054
Description
N-methyl-D-aspartate (NMDA) receptors are a class of ionotropic glutamate receptors. NMDA receptor channel has been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDA receptor channels are heteromers composed of three different subunits: NR1 (GRIN1), NR2 (GRIN2A, GRIN2B, GRIN2C, or GRIN2D) and NR3 (GRIN3A or GRIN3B). The NR2 subunit acts as the agonist binding site for glutamate. This receptor is the predominant excitatory neurotransmitter receptor in the mammalian brain. [provided by RefSeq
Sequence: C-KDAHEKDDFHHLSSpecifications
GRIN2B | |
Polyclonal | |
Unconjugated | |
ELISA (1:16000) Western Blot (2-4 ug/mL) The optimal working dilution should be determined by the end user. | |
GRIN2B | |
MGC142178/MGC142180/NMDAR2B/NR2B/hNR3 | |
Goat | |
Antigen affinity purification | |
RUO | |
2904 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of GRIN2B. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_000825.1 | |
GRIN2B | |
A synthetic peptide corresponding to human GRIN2B. | |
100 μg | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only