Learn More
GRIPAP1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of GRIPAP1.
Supplier: Abnova Corporation PAB6631
Description
GRASP1 is a neuron-specific guanine nucleotide exchange factor for the Ras family of small G proteins (RasGEF) and is associated with the GRIP/AMPA receptor complex in brain (Ye et al., 2000 [PubMed 10896157]).[supplied by OMIM
Sequence: LREQTTGLAAELQ-CSpecifications
GRIPAP1 | |
Polyclonal | |
Unconjugated | |
ELISA (1:32000) The optimal working dilution should be determined by the end user. | |
GRIPAP1 | |
DKFZp434P0630/GRASP-1/KIAA1167/MGC126593/MGC126595 | |
Goat | |
Antigen affinity purification | |
RUO | |
56850 | |
Human | |
Liquid |
ELISA | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of GRIPAP1. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_064522.3;NP_997555.1 | |
GRIPAP1 | |
A synthetic peptide corresponding to human GRIPAP1. | |
100 μg | |
Primary | |
This antibody is expected to recognize both reported isoforms (as represented by NP_064522.3, NP_997555.1) | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only