Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gstm1, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89116760 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-760 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-760 Supplier Abnova Corporation Supplier No. PAB13600
Add to Cart
Add to Cart

Goat polyclonal antibody raised against native Gstm1.

Specifications

Antigen Gstm1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against native Gstm1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In purified antiserum
Gene Gstm1
Gene Alias GSTA3
Gene Symbols Gstm1
Host Species Goat
Immunogen Native purified Gstm1 from rat liver.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24423
Test Specificity Specific for the GST mu form (in rat and human) and does not cross-react with alpha or pi class GST's. Reactive with 0.1ug of purified rat and /or human GST Yb1. It reacts strongly with GST Yb1 and Yb2 for both rodents and human (M1-1, M2-2).
Target Species Human, Rat
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.