Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTF2IRD1, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89132933 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-933 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-933 Supplier Abnova Corporation Supplier No. PAB7077
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of GTF2IRD1.

The protein encoded by this gene contains five GTF2I-like repeats and each repeat possesses a potential helix-loop-helix (HLH) motif. It may have the ability to interact with other HLH-proteins and function as a transcription factor or as a positive transcriptional regulator under the control of Retinoblastoma protein. This gene is deleted in Williams-Beuren syndrome, a multisystem developmental disorder caused by deletion of multiple genes at 7q11.23. Alternative splicing of this gene generates at least 2 transcript variants. [provided by RefSeq

Sequence: C-NKFTKDTTKLEPAS

Specifications

Antigen GTF2IRD1
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of GTF2IRD1.
Dilution ELISA (1:64000) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene GTF2IRD1
Gene Accession No. NP_057412.1;NP_005676.2
Gene Alias BEN/CREAM1/GTF3/MUSTRD1/RBAP2/WBS/WBSCR11/WBSCR12/hMusTRD1alpha1
Gene Symbols GTF2IRD1
Host Species Goat
Immunogen A synthetic peptide corresponding to human GTF2IRD1.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9569
Test Specificity This antibody is expected to recognize both reported isoforms (NP_057412.1 and NP_005676.2).
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.