Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hepatitis B surface antigen (ad/ay), Goat, Polyclonal Antibody, Abnova™

Catalog No. 89137766 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-766 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-766 Supplier Abnova Corporation Supplier No. PAB13968
Add to Cart
Add to Cart

Goat polyclonal antibody raised against hepatitis B surface antigen (ad/ay).

Specifications

Antigen hepatitis B surface antigen (ad/ay)
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against hepatitis B surface antigen (ad/ay).
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.2 (0.09% sodium azide)
Host Species Goat
Immunogen Mixture of subtypes ad and ay hepatitis B surface antigen purified from human serum.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Monospecific to purified surface antigen HBsAg.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.