Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIP1R, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89119475 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-119-475 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-119-475 Supplier Abnova Corporation Supplier No. PAB17811
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of HIP1R.

Sequence: C-RQDHQLDKKDGIYP

Specifications

Antigen HIP1R
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of HIP1R.
Dilution ELISA (1:32000) The optimal working dilution should be determined by the end user.
Formulation In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
Gene HIP1R
Gene Alias FLJ14000/FLJ27022/HIP12/HIP3/ILWEQ/KIAA0655/MGC47513
Gene Symbols HIP1R
Host Species Goat
Immunogen A synthetic peptide corresponding to human HIP1R.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9026
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.