Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIV-1, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89137778 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-778 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-778 Supplier Abnova Corporation Supplier No. PAB13983
Add to Cart
Add to Cart

Goat polyclonal antibody raised against HIV-1.

Specifications

Antigen HIV-1
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against HIV-1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.2 (0.09% sodium azide)
Host Species Goat
Immunogen Human isolate, confirmed.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Purified virions, disrupted. Reacts with the whole virus. It reacts with all the major viral proteins including p17, p24, gp41, p53, p64, gp120 and gp160. Does not react with T4+human lymphocytes.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.