Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIVEP2, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89119492 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-119-492 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-119-492 Supplier Abnova Corporation Supplier No. PAB17829
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of HIVEP2.

Members of the ZAS family, such as ZAS2 (HIVEP2), are large proteins that contain a ZAS domain, a modular protein structure consisting of a pair of C2H2 zinc fingers with an acidic-rich region and a serine/threonine-rich sequence. These proteins bind specific DNA sequences, including the kappa-B motif (GGGACTTTCC), in the promoters and enhancer regions of several genes and viruses, including human immunodeficiency virus (HIV). ZAS genes span more than 150kb and contain at least 10 exons, one of which is longer than 5.5kb (Allen and Wu, 2004).[supplied by OMIM

Sequence: C-TEESKDPSSEKSQ

Specifications

Antigen HIVEP2
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of HIVEP2.
Dilution ELISA (1:1000) The optimal working dilution should be determined by the end user.
Formulation In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
Gene HIVEP2
Gene Alias HIV-EP2/MBP-2/MIBP1/SHN2/ZAS2/ZNF40B
Gene Symbols HIVEP2
Host Species Goat
Immunogen A synthetic peptide corresponding to human HIVEP2.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3097
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.