Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HNF4A, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89132670 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-670 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-670 Supplier Abnova Corporation Supplier No. PAB6768
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of HNF4A.

The protein encoded by this gene is a nuclear transcription factor which binds DNA as a homodimer. The encoded protein controls the expression of several genes, including hepatocyte nuclear factor 1 alpha, a transcription factor which regulates the expression of several hepatic genes. This gene may play a role in development of the liver, kidney, and intestines. Mutations in this gene have been associated with monogenic autosomal dominant noninsulin-dependent diabetes mellitus type I. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq

Sequence: RLSKTLVDMDMADY-C

Specifications

Antigen HNF4A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of HNF4A.
Dilution ELISA (1:32000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene HNF4A
Gene Alias FLJ39654/HNF4/HNF4a7/HNF4a8/HNF4a9/MODY/MODY1/NR2A1/NR2A21/TCF/TCF14
Gene Symbols HNF4A
Host Species Goat
Immunogen A synthetic peptide corresponding to human HNF4A.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3172
Test Specificity This antibody is expected to recognize the reported isoforms a, b and c (NP_849180.1, NP_000448.3, NP_849181.1 resp.).
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.