Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSD11B1 Goat anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89132632 Shop All Abnova Corporation Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-632 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. 89-132-632 missing translation for 'mfr' Abnova Corporation missing translation for 'supplierNo' PAB6723
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of HSD11B1.

The protein encoded by this gene is a microsomal enzyme that catalyzes the conversion of the stress hormone cortisol to the inactive metabolite cortisone. In addition, the encoded protein can catalyze the reverse reaction, the conversion of cortisone to cortisol. Too much cortisol can lead to central obesity, and a particular variation in this gene has been associated with obesity and insulin resistance in children. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq

Sequence: CTSYNMDRFINK

Specifications

Antigen HSD11B1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of HSD11B1.
Dilution ELISA (1:4000) Western Blot (0.3-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene HSD11B1
Gene Alias 11-DH/11-beta-HSD1/HDL/HSD11/HSD11B/HSD11L/MGC13539/SDR26C1
Gene Symbols HSD11B1
Host Species Goat
Immunogen A synthetic peptide corresponding to human HSD11B1.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3290
Test Specificity This antibody is expected to recognize human HSD11B1 protein. Both NP_005516.1 and NP_861420.1 variants encode the same protein.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.