Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HTR1A, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of Htr1a.
Supplier: Abnova Corporation PAB19759
Description
Sequence: CERKNERTAEAKRKSpecifications
HTR1A | |
Polyclonal | |
Unconjugated | |
ELISA (1:128000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
Htr1a | |
Htr1a | |
A synthetic peptide correspoding to internal region of mouse Htr1a. | |
100 μg | |
Primary | |
Mouse | |
Liquid |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of Htr1a. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
Gpcr18 | |
Goat | |
Antigen affinity purification | |
RUO | |
15550 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction